Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibody detects endogenous levels of total BRI3B.
BRI3BP (BRI3 Binding Protein) is a Protein Coding gene located in mitochondrion. Involved in tumorigenesis and may function by stabilizing p53/TP53.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BRI3-binding protein; BRI3BP; Cervical cancer 1 proto-oncogene-binding protein KG19; cervical cancer oncogene binding protein; HCCRBP-1; I3-binding protein
Gene Aliases: 2410150I18Rik; AI841257; AW742481; BNAS1; BRI3BP; HCCR-1; HCCR-2; HCCRBP-1; KG19
UniProt ID: (Human) Q8WY22, (Mouse) Q8BXV2
Entrez Gene ID: (Human) 140707, (Mouse) 76809
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support