Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MSSMWSEYT IGGVKIYFPY KAYPSQLAMM NSILRGLNSK QHCLLESPTG SGKSLALLCS ALAWQQSLSG KPADEGVSEK AEVQLSCCCA CHSKDFTNND MNQGTSRHFN YPSTPPSERN GTSSTCQDSP EKTTLAAKLS AKKQASIYRD ENDDFQVEKK RIRPLETTQQ IRKRHCFGTE VHNLDAKVDS GKTVKLNSPL EKINSFSPQK PPGHCSRCCC STKQGNSQES SNTIKKDHTG KSKIPKIYFG TRTHKQIAQI TRELRRTAYS GVPMTILSSR DHTCVHPEVV GNFNRNEKCM ELLDGKNGKS CYFYHGVHKI SDQHTLQTFQ GMCKAWDIEE LVS (1-342 aa encoded by BC101472)
LIN54 is a component of the LIN, or DREAM, complex, an essential regulator of cell cycle genes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP-dependent RNA helicase BRIP1; BACH 1; BACH1 BRIP1; BRCA1-associated C-terminal helicase 1; BRCA1-binding helicase-like protein BACH1; BRCA1-interacting protein 1; BRCA1-interacting protein C-terminal helicase 1; BRCA1/BRCA2-associated helicase 1; BRIP 1; DNA 5'-3' helicase FANCJ; FANCJ protein; Fanconi anemia group J protein; Fanconi anemia group J protein homolog; FLJ90232; MGC126521; MGC126523
Gene Aliases: 3110009N10Rik; 8030460J03Rik; BACH1; BRIP1; FACJ; FANCJ; OF
UniProt ID: (Human) Q9BX63, (Mouse) Q5SXJ3
Entrez Gene ID: (Human) 83990, (Mouse) 237911
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support