Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
BRN3A is a class IV POU domain-containing transcription factor highly expressed in the developing sensory nervous system and in cells of the B- and T-lymphocytic lineages.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Brain-3A; Brain-specific homeobox/POU domain protein 3A; Brn-3.0; brn-3A; FLJ13449; Homeobox/POU domain protein RDC-1; Oct-T1; POU domain, class 4, transcription factor 1
Gene Aliases: Brn-3; Brn-3.0; brn-3A; Brn3; Brn3.0; BRN3A; E130119J07Rik; Oct-T1; POU4F1; RDC-1; RDC1
UniProt ID: (Human) Q01851, (Mouse) P17208
Entrez Gene ID: (Human) 5457, (Mouse) 18996, (Rat) 114503
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support