Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MGVDFDVKTF CHNLRATKPP YECPVETCRK VYKSYSGIEY HLYHYDHDNP PPPQQTPLRK HKKKGRQSRP ANKQSPSPSE VSQSPGREVM SYAQAQRMVE VDLHGRVHRI SIFDNLDVVS EDEEAPEEAP ENGSNKENTE TPAATPKSGK HKNKEKRKDS NHHHHHNVSA STTPKLPEVV YRELEQDTPD APPRPTSYYR YIEKSAEELD EEVEYDMDEE DYIWLDIMNE RRKTEGVSPI PQEIFEYLMD RLEKESYFES HNKGDPNALV DEDAVCCICN DGECQNSNVI LFCDMCNLAV HQECYGVPYI PEGQWLCRRC LQSPSRAVDC ALCPNKGGAF KQTDDGRWAH VVCALWIPEV CFANTVFLEP IDSIEHIPPA RWKLTCYICK QRGSGACIQC HKANCYTAFH VTCAQQAGLY MKMEPVRETG ANGTSFSVRK TAYCDIHTPP GSARRLPALS HSEGEEDEDE EEDEGKGWSS EKVKKAKAKS RIKMKKARKI LAEKRAAAPV VSVPCIPPHR LSKITNRLTI QRKSQFMQRL HSYWTLKRQS RNGVPLLRRL QTHLQSQRNC DQVGRDSEDK NWALKEQLKS WQRLRHDLER ARLLVELIRK REKLKRETIK VQQIAMEMQL TPFLILLRKT LEQLQEKDTG NIFSEPVPLS EVTELDEVPD YLDHIKKPMD FFTMKQNLEA YRYLNFDDFE EDFNLIVSNC LKYNAKDTIF YRAAVRLREQ GGAVLRQARR QAEKMGIDFE TGMHIPHSLA GDEATHHTED AAEEERLVLL ENQKHLPVEE QLKLLLERLD EVNASKQSVG RSRRAKMIKK EMTALRRKLA HQRETGRDGP ERHGPSSRGS LTPHPAACDK DGQTDSAAEE SSSQETSKGL GPNMSSTPAH EVGRRTSVLF SKKNPKTAGP PKRPGRPPKN RESQMTPSHG GSPVGPPQLP IMSSLRQRKR GRSPRPSSSS DSDSDKSTED PPMDLPANGF SGGNQPVKKS FLVYRNDCSL PRSSSDSESS SSSSSSAASD RTSTTPSKQG RGKPSFSRGT FPEDSSEDTS GTENEAYSVG TGRGVGHSMV RKSLGRGAGW LSEDEDSPLD ALDLVWAKCR GYPSYPALII DPKMPREGMF HHGVPIPVPP LEVLKLGEQM TQEAREHLYL VLFFDNKRTW QWLPRTKLVP LGVNQDLDKE KMLEGRKSNI RKSVQIAYHR ALQHRSKVQG EQSSETSDSD
BRPF1 (peregrin) is a component of the MOZ/MORF complex. This complex has histone H3 acetyltransferase activity. BRPF1 preferentially mediates histone H3-K23 acetylation and positively regulates the transcription of RUNX1 and RUNX2. Mutations affecting the gene can result in intellectual developmental disorder with dysmorphic facies and ptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Bromodomain and PHD finger-containing protein 1; bromodomain-containing protein, 140kD; Peregrin; Protein Br140
Gene Aliases: BR140; BRPF1
UniProt ID: (Human) P55201
Entrez Gene ID: (Human) 7862
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support