Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Sequence of this protein is as follows: MTAAPGGSAQ PAGPGPRLGF STADSGVGMS GLNPGPAVPM KDHDAIKLFV GQIPRGLDEQ DLKPLFEEFG RIYELTVLKD RLTGLHKGCA FLTYCARDSA LKAQSALHEQ KTLPGMNRPI QVKPAASEGR GEDRKLFVGM LGKQQGEEDV RRLFQPFGHI EECTVLRSPD GTSKGCAFVK FGSQGEAQAA IRGLHGSRTM AGASSSLVVK LADTDRERAL RRMQQMAGHL GAFHPAPLPL GACGAYTTAI LQHQAALLAA AQGPGLGPVA AVAAQMQHVA AFSLVAAPLL PAAAANSPPG SGPGTLPGLP APIGVNGFGP LTPQTNGQPG SDTLYNNGLS PYPAQSPGVA DPLQQAYAGM HHYAAAYPSA YAPVSTAFPQ QPSALPQQQR EGPEGCNLFI YHLPQEFGDA ELIQTFLPFG AVVSAKVFVD RATNQSKCFG FVSFDNPTSA QTAIQAMNGF QIGMKRLKAQ LKRPKDANRP Y
RNA-binding protein implicated in the regulation of pre-mRNA alternative splicing. Mediates exon inclusion and/or exclusion in pre-mRNA that are subject to tissue-specific and developmentally regulated alternative splicing. Specifically activates exon 5 inclusion of TNNT2 in a muscle-specific splicing enhancer (MSE)-dependent manner. Promotes also exon exclusion of INSR pre-mRNA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Bruno -like 6, RNA binding protein; bruno-like 6, RNA binding protein; Bruno-like protein 6; CELF-6; CUG-BP and ETR-3 like factor 6; CUG-BP- and ETR-3-like factor 6; CUGBP Elav-like family member 6; RNA-binding protein BRUNOL-6
Gene Aliases: BRUNOL6; CELF6
UniProt ID: (Human) Q96J87
Entrez Gene ID: (Human) 60677
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support