Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSAEGAEPGP GSGSGPGPGP LCPEHGQALS WFCGSERRPV CAACAGLGGR CRGHRIRRAE ERAEELRNKI VDQCERLQLQ SAAITKYVAD VLPGKNQRAV SMASAARELV IQRLSLVRSL CESEEQRLLE QVHGEEERAH QSILTQRVHW AEALQKLDTI RTGLVGMLTH LDDLQLIQKE QEIFERHRGH TDR
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B box and SPRY domain-containing protein; B-box and SPRY-domain containing protein; zetin 1
Gene Aliases: BSPRY
UniProt ID: (Human) Q5W0U4
Entrez Gene ID: (Human) 54836
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support