Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • BTBD12 Antibodies

          Abnova

          BTBD12 Polyclonal Antibody, MaxPab™

          View all (7) BTBD12 antibodies
          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite BTBD12 Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (1)
          BTBD12 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          BTBD12 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          BTBD12 Antibody (H00084464-B01P) in WB

          Western Blot analysis of BTBD12 expression in transfected 293T cell line (H00084464-T02) by BTBD12 MaxPab polyclonal antibody.

          Lane 1: BTBD12 transfected lysate(126.61 KDa).
          Lane 2: Non-transfected lysate.
          {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          BTBD12 Antibody in Western Blot (WB)

          Product Details

          H00084464-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          BTBD12 (AAH36335.1, 1 a.a. approximately 1151 a.a) full-length human protein.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MVNNPHLSDV QFQTDSGEVL YAHKFVLYAR CPLLIQYVNN EGFSAVEDGV LTQRVLLGDV STEAARTFLH YLYTADTGLP PGLSSELSSL AHRFGVSELV HLCEQVPIAT DSEGKPWEEK EAENCESRAE NFQELLRSMW ADEEEEAETL LKSKDHEEDQ ENVNEAEMEE IYEFAATQRK LLQEERAAGA GEDADWLEGG SPVSGQLLAG VQVQKQWDKV EEMEPLEPGR DEAATTWEKM GQCALPPPQG QHSGARGAEA PEQEAPEEAL GHSSCSSPSR DCQAERKEGS LPHSDDAGDY EQLFSSTQGE ISEPSQITSE PEEQSGAVRE RGLEVSHRLA PWQASPPHPC RFLLGPPQGG SPRGSHHTSG SSLSTPRSRG GTSQVGSPTL LSPAVPSKQK RDRSILTLSK EPGHQKGKER RSVLECRNKG VLMFPEKSLS IDLTQSNPDH SSSRSQKSSS KLNEEDEVIL LLDSDEELEL EQTKMKSISS DPLEEKKALE ISPRSCELFS IIDVDADQEP SQSPPRSEAV LQQEDEGALP ENRGSLGRRG APWLFCDRES SPSEASTTDT SWLVPATPLA SRSRDCSSQT QISSLRSGLA VQAVTQHTPR ASVGNREGNE VAQKFSVIRP QTPPPQTPSS CLTPVSPGTS DGRRQGHRSP SRPHPGGHPH SSPLAPHPIS GDRAHFSRRF LKHSPPGPSF LNQTPAGEVV EVGDSDDEQE VASHQANRSP PLDSDPPIPI DDCCWHMEPL SPIPIDHWNL ERTGPLSTSS PSRRMNEAAD SRDCRSPGLL DTTPIRGSCT TQRKLQEKSS GAGSLGNSRP SFLNSALWDV WDGEEQRPPE TPPPAQMPSA GGAQKPEGLE TPKGANRKKN LPPKVPITPM PQYSIMETPV LKKELDRFGV RPLPKRQMVL KLKEIFQYTH QTLDSDSEDE SQSSQPLLQA PHCQTLASQT YKPSRAGVHA QQEATTGPGA HRPKGPAKTK GPRHQRKHHE SITPPSRSPT KEAPPGLNDD AQIPASQESV ATSVDGSDSS LSSQSSSSCE FGAAFESAGE EEGEGEVSAS QAAVQAADTD EALRCYIRSK PALYQKVLLY QPFELRELQA ELRQNGLRVS SRRLLDFLDT HCITFTTAAT RREKLQGRRR QPRGKKKVER N

          Target Information

          This gene encodes a structure-specific endonuclease subunit. The encoded protein contains a central BTB domain and it forms a multiprotein complex with the ERCC4(XPF)-ERCC1, MUS81-EME1, and SLX1 endonucleases, and also associates with MSH2/MSH3 mismatch repair complex, telomere binding complex TERF2(TRF2)-TERF2IP(RAP1), the protein kinase PLK1 and the uncharacterized protein C20orf94. The multiprotein complex is required for repair of specific types of DNA lesions and is critical for cellular responses to replication fork failure. The encoded protein acts as a docking platform for the assembly of multiple structure-specific endonucleases.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: BTB (POZ) domain containing 12; BTB/POZ domain-containing protein 12; SLX4 structure-specific endonuclease subunit homolog; Structure-specific endonuclease subunit SLX4

          View more View less

          Gene Aliases: BTBD12; FANCP; KIAA1784; KIAA1987; MUS312; SLX4

          View more View less

          UniProt ID: (Human) Q8IY92

          View more View less

          Entrez Gene ID: (Human) 84464

          View more View less

          Function(s)
          endodeoxyribonuclease activity protein binding enzyme activator activity crossover junction endodeoxyribonuclease activity 5'-flap endonuclease activity 3'-flap endonuclease activity
          Process(es)
          double-strand break repair via homologous recombination DNA replication DNA repair nucleotide-excision repair DNA double-strand break processing involved in repair via single-strand annealing interstrand cross-link repair positive regulation of catalytic activity response to intra-S DNA damage checkpoint signaling t-circle formation positive regulation of t-circle formation
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-9267x:80/100.66.134.224:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0