Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: FVVGPPGATR TVPAHKYVLA VGSSVFYAMF YGDLAEVKSE IHIPDVEPAA
Sequence homology: Cow: 86%; Guinea Pig: 79%; Horse: 79%; Human: 100%; Rabbit: 79%; Zebrafish: 79%
Adapter protein for the CUL3 E3 ubiquitin-protein ligase complex. Involved in late neuronal development and muscle formation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BTB (POZ) domain containing 6; BTB domain protein BDPL; BTB/POZ domain-containing protein 6; glucocorticoid receptor AF-1 coactivator-1; Lens BTB domain protein
Gene Aliases: BDPL; BTBD6
UniProt ID: (Human) Q96KE9
Entrez Gene ID: (Human) 90135
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support