Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MCRALLYGGM RESQPEAEIP LQDTTAEAFT MLLKYIYTGR ATLTDEKEEV LLDFLSLAHK YGFPELEDST SEYLCTILNI QNVCMTFDVA SLYSLPKLTC MCCMFMDRNA QEVLSSEGFL SLSKTALLNI VLRDSFAAPE KDIFLALLNW CKHNSKENHA EIMQAVRLPL MSLTELLNVV RPSGLLSPDA ILDAIKVRSE SRDMDLNYRG MLIPEENIAT MKYGAQVVKG ELKSALLDGD TQNYDLDHGF SRHPIDDDCR SGIEIKLGQP SIINHIRILL WDRDSRSYSY FIEVSMDELD WVRVIDHSQY LCRSWQKLYF PARVCSGDGV SLWCPLWSRT PELKQSSLLG LPKCRYIRIV GTHNTVNKIF HIVAFECMFT NKTFTLEKGL IVPMENVATI ADCASVIEGV SRSRNALLNG DTKNYDWDSG YTCHQLGSGA IVVQLAQPYM IGSIRLLLWD CDDRSYSYYV EVSTNQQQWT MVADRTKVSC KSWQSVTFER QPASFIRIVG THNTANEVFH CVHFECPEQQ SSQKEENSEE SGTGDTSLAG QQLDSHALRA PSGSSLPSSP GSNSRSPNRQ HQ
This locus encodes a BTB/POZ domain-containing protein. This domain is known to be involved in protein-protein interactions. Polymorphisms at this locus have been reported to be associated with susceptibility to Restless Legs Syndrome and may also be associated with Tourette Syndrome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BTB (POZ) domain containing 9; BTB/POZ domain-containing protein 9
Gene Aliases: BTBD9; dJ322I12.1; KIAA1880
UniProt ID: (Human) Q96Q07
Entrez Gene ID: (Human) 114781
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support