Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Sequence of this protein is as follows: MDTPENVLQM LEAHMQSYKG NDPLGVWERY IQWVEENFPE NKEYLITLLE HLMKEFLDKK KYHNDPRFIS YCLKFAEYNS DLRQFFEFLY NHGIGTLSSP LYIAWAGHLE AQGELQHASA VLQRGIQNQA EPREFLQQQY RLFQTRLTET HLPAQARTSE PLHNVQVLNQ MITSKSNPGN NMACISKNQG SELSGVISSA CDKESNMERR VITISKSEYS VHSSLASKVD VEQVVMYCKE KLIRGESEFS FEELRAQKYN QRRKHEQWVN EDRHYMKRKE ANAFEEQLLK QKMDELHKKL HQVVETSHED LPASQERSEV NPARMGPSVG SQQELRAPCL PVTYQQTPVN MEKNPREAPP VVPPLANAIS AALVSPATSQ SIAPPVPLKA QTVTDSMFAV ASKDAGCVNK STHEFKPQSG AEIKEGCETH KVANTSSFHT TPNTSLGMVQ ATPSKVQPSP TVHTKEALGF IMNMFQAPTL PDISDDKDEW QSLDQNEDAF EAQFQKNVRS SGAWGVNKII SSLSSAFHVF EDGNKENYGL PQPKNKPTGA RTFGERSVSR LPSKPKEEVP HAEEFLDDST VWGIRCNKTL APSPKSPGDF TSAAQLASTP FHKLPVESVH ILEDKENVVA KQCTQATLDS CEENMVVPSR DGKFSPIQEK SPKQALSSHM YSASLLRLSQ PAAGGVLTCE AELGVEACRL TDTDAAIAED PPDAIAGLQA EWMQMSSLGT VDAPNFIVGN PWDDKLIFKL LSGLSKPVSS YPNTFEWQCK LPAIKPKTEF QLGSKLVYVH HLLGEGAFAQ VYEATQGDLN DAKNKQKFVL KVQKPANPWE FYIGTQLMER LKPSMQHMFM KFYSAHLFQN GSVLVGELYS YGTLLNAINL YKNTPEKVMP QGLVISFAMR MLYMIEQVHD CEIIHGDIKP DNFILGNGFL EQDDEDDLSA GLALIDLGQS IDMKLFPKGT IFTAKCETSG FQCVEMLSNK PWNYQIDYFG VAATVYCMLF GTYMKVKNEG GECKPEGLFR RLPHLDMWNE FFHVMLNIPD CHHLPSLDLL RQKLKKVFQQ HYTNKIRALR NRLIVLLLEC KRSRK
BUB1A is a kinase involved in spindle checkpoint function. The kinase functions in part by phosphorylating a member of the miotic checkpoint complex and activating the spindle checkpoint.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BUB1 budding uninhibited by benzimidazoles 1 homolog; BUB1, mitotic checkpoint serine/threonine kinase; BUB1A; budding uninhibited by benzimidazoles 1 homolog; hBUB1; Mitotic checkpoint serine/threonine-protein kinase BUB1; mitotic spindle checkpoint kinase; putative serine/threonine-protein kinase
Gene Aliases: BUB1; BUB1A; BUB1L; hBUB1
UniProt ID: (Human) O43683
Entrez Gene ID: (Human) 699
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support