Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • Blimp-1 Antibodies

          Invitrogen

          Blimp-1 Monoclonal Antibody (6D3), eBioscience™

          Advanced Verification
          28 Published Figures
          26 References
          View all (39) Blimp-1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Blimp-1 Monoclonal Antibody (6D3), eBioscience™

          • Antibody Testing Data (3)
          • Published Figures (28)
          • Advanced Verification (1)
          Blimp-1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Blimp-1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 32

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Blimp-1 Antibody (14-5963-82) in IHC (P)

          Immunohistochemistry on formalin-fixed paraffin embedded human tonsil tissue with citrate buffer antigen retrieval, using 10 µg/mL of Anti-Human/Mouse Blimp1 Purified or 10 µg/mL of Rat IgG2a kappa Isotype Control Purified (Product # 14-4321-82) followed by Anti-Rat IgG Biotin, and DAB visualization. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Blimp-1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Blimp-1 Antibody in Western Blot (WB)
          Blimp-1 Antibody in Flow Cytometry (Flow)
          Blimp-1 Antibody in Immunocytochemistry (ICC/IF)
          Blimp-1 Antibody in Immunocytochemistry, Immunohistochemistry (ICC/IF, IHC)
          Blimp-1 Antibody in Immunocytochemistry, Immunohistochemistry (Frozen) (ICC/IF, IHC (F))
          Blimp-1 Antibody in Immunocytochemistry (ICC/IF)
          Blimp-1 Antibody in Immunocytochemistry (ICC/IF)
          Blimp-1 Antibody in Immunocytochemistry (ICC/IF)
          Blimp-1 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Blimp-1 Antibody in Immunohistochemistry, Immunohistochemistry (Paraffin) (IHC, IHC (P))
          Blimp-1 Antibody in Immunohistochemistry (IHC)
          Blimp-1 Antibody in Immunohistochemistry (IHC)
          Blimp-1 Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Blimp-1 Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Blimp-1 Antibody in Immunohistochemistry (IHC)
          Blimp-1 Antibody in Immunohistochemistry (IHC)
          Blimp-1 Antibody in Immunohistochemistry (IHC)
          Blimp-1 Antibody in Immunohistochemistry (IHC)
          Blimp-1 Antibody in Immunohistochemistry (IHC)
          Blimp-1 Antibody in Western Blot (WB)
          Blimp-1 Antibody in Western Blot (WB)
          Blimp-1 Antibody in Western Blot (WB)
          Blimp-1 Antibody in Western Blot (WB)
          Blimp-1 Antibody in Western Blot (WB)
          Blimp-1 Antibody in Flow Cytometry (Flow)
          Blimp-1 Antibody in ChIP Assay (ChIP)
          Blimp-1 Antibody in ChIP Assay (ChIP)
          Blimp-1 Antibody in ChIP Assay (ChIP)
          Blimp-1 Antibody in ChIP Assay (ChIP)
          Blimp-1 Antibody in ChIP Assay (ChIP)
          Blimp-1 Antibody

          Product Details

          14-5963-82

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-10 µg/mL
          View 8 publications 8 publications

          Immunohistochemistry (IHC)

          -
          View 8 publications 8 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          10 µg/mL
          View 2 publications 2 publications

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          -
          View 4 publications 4 publications

          Flow Cytometry (Flow)

          (0.5 µg)/test)
          View 1 publication 1 publication

          ChIP assay (ChIP)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Human, Mouse

          Published species

          Human, Mouse

          Host/Isotype

          Rat / IgG2a

          Class

          Monoclonal

          Type

          Antibody

          Clone

          6D3

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_1907437

          Product Specific Information

          Description: This 6D3 monoclonal antibody reacts with B lymphocyte-induced maturation protein-1 (Blimp-1), a transcriptional repressor expressed in B and T cells. This approximately 90-kDa protein contains five consecutive Kruppel-like zinc finger domains by which it binds DNA. In B cells, Blimp-1 promotes terminal differentiation into antibody-secreting plasma cells during a primary immune response. In addition, Blimp-1 has been implicated in germinal center formation in response to T-dependent and T-independent antigens, as well as in some diffuse large B cell lymphomas. Expressed in CD4+ and CD8+ effector/memory T cells and FoxP3+ regulatory T cells, reports have suggested possible roles for Blimp-1 in T cell homeostasis and suppression of IL-2 production. Moreover, by suppressing Th1 differentiation, Blimp-1 plays a role in T helper cell differentiation. Finally, Blimp-1 has been demonstrated to play a significant role in the terminal differentiation of effector and effector memory CD8+ T cells during viral infection.

          Applications Reported: This 6D3 antibody has been reported for use in immunoblotting (WB) and immunohistology staining of paraffin-embedded tissue sections.

          Applications Tested: This 6D3 antibody has been tested by western blot of reduced A20 cell lysates. For western blottiing, this antibody can be used at 1-10 µg/mL. Moreover, this antibody was tested by immunohistochemistry of formalin-fixed paraffin embedded human tonsil tissue. Antigen retrieval was performed using 10mM citrate buffer at pH 6.0. For immunohistochemistry, this antibody can be used at 10 µg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Purity: Greater than 90%, as determined by SDS-PAGE.

          Aggregation: Less than 10%, as determined by HPLC.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          Blimp-1 (B lymphocyte maturation protein-1) is a nuclear protein with five zinc fingers in its carboxy terminus. It is a transcriptional repressor that is both required and sufficient to trigger terminal differentiation of B lymphocytes and monocyte/macrophages. Blimp-1 is expressed in, and may be required for terminally differentiated epithelial cells, retinal neurons and other lineage cells. It has also been implicated in the repression of beta-interferon (IFN-beta) and c-myc gene expression.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: B lymphocyte-induced maturation protein 1; B-lymphocyte-induced maturation protein 1; Beta-interferon gene positive regulatory domain I-binding factor; beta-interferon gene positive-regulatory domain I binding factor; BLIMP 1; BLIMP-1; MGC118922; MGC118923; MGC118924; MGC118925; OTTHUMP00000016918; OTTHUMP00000202173; Positive regulatory domain I-binding factor 1; PR domain containing 1, with ZNF domain; PR domain zinc finger protein 1; PR domain-containing protein 1; PRDI BF1; PRDI-BF1; PRDI-binding factor-1; RP1-134E15.1

          View more View less

          Gene Aliases: b2b1765Clo; Blimp-1; BLIMP1; PRDI-BF1; PRDM1; ZNFPR1A1

          View more View less

          UniProt ID: (Human) O75626, (Mouse) Q60636

          View more View less

          Entrez Gene ID: (Human) 639, (Mouse) 12142

          View more View less

          Function(s)
          RNA polymerase II core promoter proximal region sequence-specific DNA binding transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding transcription factor activity, sequence-specific DNA binding protein binding methyltransferase activity histone deacetylase binding metal ion binding promoter-specific chromatin binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter morphogenesis of a branching structure maternal placenta development heart valve development ventricular septum development transcription, DNA-templated germ cell development post-embryonic development positive regulation of gene expression negative regulation of B cell proliferation negative regulation of lipopolysaccharide-mediated signaling pathway methylation regulation of natural killer cell differentiation regulation of extrathymic T cell differentiation aorta development eye photoreceptor cell development cell fate commitment positive regulation of B cell differentiation artery morphogenesis regulation of NK T cell differentiation intestinal epithelial cell development trophoblast giant cell differentiation coronary vasculature development sebum secreting cell proliferation
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-67f4cd694f-jv9gd:80/100.66.135.11:80.
          git-commit: 961b7666e63bcc533d6299fdd217d6a70bd96cf1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0