Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAQVLIVGAG MTGSLCAALL RRQTSGPLYL AVWDKADDSG GRMTTACSPH NPQCTADLGA QYITCTPHYA KKHQRFYDEL LAYGVLRPLS SPIEGMVMKE GDCNFVAPQG ISSIIKHYLK ESGAEVYFRH RVTQINLRDD KWEVSKQTGS PEQFDLIVLT MPVPEILQLQ GDITTLISEC QRQQLEAVSY SSRYALGLFY EAGTKIDVPW AGQYITSNPC IRFVSIDNKK RNIESSEIGP SLVIHTTVPF GVTYLEHSIE DVQELVFQQL ENILPGLPQP IATKCQKWRH SQVTNAAANC PGQMTLHHKP FLACGGDGFT QSNFDGCITS ALCVLEALKN YI
Renalase is a flavin adenine dinucleotide-dependent amine oxidase that is secreted into the blood from the kidney.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha-NAD(P)H oxidase/anomerase; FLJ11218; HGNC:25641; MAO-C; Monoamine oxidase-C; Renalase; renalase, FAD-dependent amine oxidase
Gene Aliases: C10orf59; RENALASE; RNLS
UniProt ID: (Human) Q5VYX0
Entrez Gene ID: (Human) 55328
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support