Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MDNCLAAAAL NGVDRRSLQR SARLALEVLE RAKRRAVDWH ALERPKGCMG VLAREAPHLE KQPAAGPQRV LPGEREERPP TLSASFRTMA EFMDYTSSQC GKYYSSVPEE GGATHVYRYH RGESKLHMCL DIGNGQRKDR KKTSLGPGGS YQISEHAPEA SQPAENISKD LYIEVYPGTY SVTVGSNDLT KKTHVVAVDS GQSVDLVFPV
Enhances NF-kappa-B transcriptional activity by regulating the nuclear localization of the NF-kappa-B subunit RELA and promoting the phosphorylation of RELA by PRKACA. Regulates the effect of the cAMP-dependent protein kinase signaling pathway on the NF-kappa-B activation cascade.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A kinase (PRKA) interacting protein 1; A-kinase-interacting protein 1; breast cancer associated gene 3; Breast cancer-associated gene 3 protein; koyt binding protein 1; koyt binding protein 2; koyt binding protein 3; PKA-interacting protein; Proline-rich protein BCA3
Gene Aliases: AKIP1; BCA3; C11orf17
UniProt ID: (Human) Q9NQ31
Entrez Gene ID: (Human) 56672
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support