Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MLVGTPNLL TLDEADATWT LIKDKVIEEH FGPNAVAVPF LSDAACYDLL GVLVKQSRPA HTRLALPGRQ GRRALKPVGP LPSLLEQAGS EGAFAHCTRE YSPNGRAERA YEETRMLDGQ PCKIRLHMGD LRKKVAFLLL PPGQVSLQQT LPWLRSTHSI YVIYQVFSCS WLQLGLTSTA REPQLLRLLR SLPVAFSCLK FSLQSKGVLG PQKPLTKDPL PHGANWVRPN LSIMPPLAPT SASADTTEAA DVPPPVPAPP TPPPQEGPED KPTRFSYKGR NPFWRGPQIL SENWLFSPRS PPPGAQGGGP RDPDGHSMSL PLLQGLSSEF DSDD (1-333 aa encoded by BC031612)
C11orf42 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Uncharacterized protein C11orf42
Gene Aliases: C11orf42
UniProt ID: (Human) Q8N5U0
Entrez Gene ID: (Human) 160298
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support