Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
This gene encodes the auxiliary subunit of the heteromeric N-terminal acetyltransferase B complex. This complex acetylates methionine residues that are followed by acidic or asparagine residues.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: liver regeneration-related protein LRRGT00164; mitochondrial distribution and morphology 20; Mitochondrial distribution and morphology protein 20; N-alpha-acetyltransferase 25, NatB auxiliary subunit; N-terminal acetyltransferase B complex subunit MDM20; N-terminal acetyltransferase B complex subunit NAA25; Nalpha acteyltransferase 25; NatB complex subunit MDM20; p120
Gene Aliases: 4833422K13Rik; A630046C11; AI450868; C12orf30; C330023M02Rik; LRRGT00164; MDM20; NAA25; NAP1; RGD1305685
UniProt ID: (Human) Q14CX7, (Mouse) Q8BWZ3
Entrez Gene ID: (Human) 80018, (Rat) 360811, (Mouse) 231713
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support