Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Adding 0.2 mL of distilled water will yield a concentration of 500 µg/mL.
Positive Control - WB: human MOLT-4 whole cell, human U251 whole cell. Flow: RT4 cell.|Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
RHNO1 plays a role in DNA damage response (DDR) signaling upon genotoxic stresses such as ionizing radiation (IR) during the S phase. Recruited to sites of DNA damage through interaction with the 9-1-1 cell-cycle checkpoint response complex and TOPBP1 in a ATR-dependent manner. Required for the progression of the G1 to S phase transition. Plays a role in the stimulation of CHEK1 phosphorylation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RAD9, HUS1, RAD1-interacting nuclear orphan protein 1; Rad9, Rad1, Hus1 interacting nuclear orphan; RAD9, RAD1, HUS1-interacting nuclear orphan protein
Gene Aliases: C12orf32; HKMT1188; RHINO; RHNO1
UniProt ID: (Human) Q9BSD3
Entrez Gene ID: (Human) 83695
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support