Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MKKTNPGPVG ETLLCQLSSY AKTELGSQTP ESSRSEASRI LDEDSWSDGE QEPITVDQTW RGDPDSEADS IDSDQEDP
Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
The C12orf41 gene, also known as CFAP70 (Cilia and Flagella Associated Protein 70), is located on chromosome 12 and encodes a protein that is essential for the proper function of cilia and flagella. The CFAP70 protein plays a critical role in the assembly and maintenance of axonemal structures, which are core components of cilia and flagella. Cilia and flagella are vital for various cellular processes, including motility, fluid movement across cell surfaces, and signal transduction. Mutations in CFAP70 have been linked to primary ciliary dyskinesia (PCD), a genetic disorder characterized by chronic respiratory tract infections, abnormal situs (organ positioning), and infertility due to defective motility of cilia and flagella. CFAP70 is also involved in the regulation of dynein arms and radial spokes, which are necessary for the effective beating of cilia and flagella. Research into CFAP70 continues to uncover its multifaceted roles in cellular function and its potential implications in ciliopathies.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: KAT8 regulatory NSL complex subunit 2; Non-specific lethal 2 homolog; NSL complex protein NSL2
Gene Aliases: C12orf41; KANSL2; NSL2
UniProt ID: (Human) Q9H9L4
Entrez Gene ID: (Human) 54934
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support