Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MVNLAAMVWR RLLRKRWVLA LVFGLSLVYF LSSTFKQEER AVRDRNLLQV HDHNQPIPWK VQFNLGNSSR PSNQCRNSIQ GKHLITDELG YVCERKDLLV NGCCNVNVPS TKQYCCDGCW PNGCCSAYEY CVSCCLQPNK QLLLERFLNR AAVAFQNLFM AVEDHFELCL AKCRTSSQSV QHENTYRDPI AKYCYGESPP ELFPA
The exact function of C12orf49 remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: SREBF pathway regulator in Golgi 1; SREBP regulating gene protein
Gene Aliases: C12orf49; SPRING; SPRING1
UniProt ID: (Human) Q9H741
Entrez Gene ID: (Human) 79794
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support