Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MAEAVLIDLF GLKLNSQKNC HQTLLKTLNA VQYHHAAKAK FLCIMCCSNI SYERDGEQDN CEIETSNGLS ALLEEFEIVS CPSMAATLYT IKQKIDEKNL SSIKVIVPRH RKTLMKAFID QLFTDVYNFE FEDLQVTFRG GLFKQSIEIN VITAQELRGI QNEIETFLRS LPALRGKLTI ITSSLIPDIF IHGFTTRTGG ISYIPTLSSF NLFSSSKRRD PKVVVQENLR RLANAAGFNV EKFYRIKTHH SNDIWIMGRK EPDSYDGITT NQRGVTIAAL GADCIPIVFA DPVKKACGVA HAGWKGTLLG VAMATVNAMI AEYGCSLEDI VVVLGPSVGP CCFTLPRESA EAFHNLHPAC VQLFDSPNPC IDIRKATRIL LEQGGILPQN IQDQNQDLNL CTSCHPDKFF SHVRDGLNFG TQIGFISIKE
This gene encodes an oxidoreductase that promotes fatty-acid oxidation, with concomitant inflammasome activation, mitochondrial and NADPH-oxidase-dependent reactive oxygen species production, and bactericidal activity of macrophages. The encoded protein forms a complex with fatty acid synthase on peroxisomes and is thought to be modulated by peroxisome proliferator-activated receptor signaling events. Naturally occurring mutations in this gene are associated with inflammatory bowel disease, Behcet's disease, leprosy, ulcerative colitis, early-onset Crohn's disease, and systemic juvenile idiopathic arthritis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adenosine deaminase LACC1; Fatty acid metabolism-immunity nexus; Guanosine phosphorylase LACC1; laccase (multicopper oxidoreductase) domain containing 1; Laccase domain-containing protein 1; Purine nucleoside phosphorylase LACC1; S-methyl-5'-thioadenosine phosphorylase LACC1
Gene Aliases: C13orf31; FAMIN; LACC1
UniProt ID: (Human) Q8IV20
Entrez Gene ID: (Human) 144811
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support