Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MRVLVGGGTG FIGTALTQLL NARGHEVTLV SRKPGPGRIT WDELAASGLP SCDAAVNLAG ENILNPLRRW NETFQKEVLG SRLETTQLLA KAITKAPQPP KAWVLVTGVA YYQPSLTAEY DEDSPGGDFD FFSNLVTKWE AAARLPGDST RQVVVRSGVV LGRGGGAMGH MLLPFRLGLG GPIGSGHQFF PWIHIGDLAG ILTHALEANH VHGVLNGVAP SSATNAEFAQ TFGAALGRRA FIPLPSAVVQ AVFGRQRAIM LLEGQKVIPR RTLATGYQYS FPELGAALKE IVA
Putative NADP-dependent oxidoreductase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Epimerase family protein SDR39U1; short chain dehydrogenase/reductase family 39U, member 1; Short-chain dehydrogenase/reductase family 39U member 1
Gene Aliases: C14orf124; HCDI; SDR39U1
UniProt ID: (Human) Q9NRG7
Entrez Gene ID: (Human) 56948
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support