Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
OriGene
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
RTRAF (C14orf166) is an RNA-binding protein involved in modulation of mRNA transcription by Polymerase II (PubMed:16950395). Component of the tRNA-splicing ligase complex and is required for tRNA ligation (PubMed:24870230). May be required for RNA transport (PubMed:24608264).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CLE; CLE7 homolog; RLL motif containing 1; RNA transcription, translation and transport factor protein
Gene Aliases: C14orf166; CGI-99; CGI99; CLE; CLE7; hCLE1; LCRP369; RLLM1; RTRAF
UniProt ID: (Human) Q9Y224
Entrez Gene ID: (Human) 51637
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support