Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSFVAYEELI KEGDTAILSL GHGAMVAVRV QRGAQTQTRH GVLRHSVDLI GRPFGSKVTC GRGGWVYVLH PTPELWTLNL PHRTQILYST DIALITMMLE LRPGSVVCES GTGSGSVSHA IIRTIAPTGH LHTVEFHQQR AEKAREEFQE HRVGRWVTVR TQDVCRSGFG VSHVADAVFL DIPSPWEAVG HAWDALKVEG GRFCSFSPCI EQVQRTCQAL AARGFSELST LEVLPQVYNV RTVSLPPPDL GTGTDGPAGS DTSPFRSGTP MKEAVGHTGY LTFATKTPG
Catalytic subunit of tRNA (adenine-N(1)-)-methyltransferase, which catalyzes the formation of N(1)-methyladenine at position 58 (m1A58) in initiator methionyl-tRNA (PubMed:16043508). Catalytic subunit of mRNA N(1)-methyltransferase complex, which mediates methylation of adenosine residues at the N(1) position of a small subset of mRNAs: N(1) methylation takes place in tRNA T-loop-like structures of mRNAs and is only present at low stoichiometries (PubMed:29107537, PubMed:29072297). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: mRNA methyladenosine-N(1)-methyltransferase catalytic subunit TRMT61A; tRNA (adenine(58)-N(1))-methyltransferase catalytic subunit TRMT61A; tRNA (adenine-N(1)-)-methyltransferase catalytic subunit TRM61; tRNA (adenine-N(1)-)-methyltransferase catalytic subunit TRMT61A; tRNA methyltransferase 61 homolog A; tRNA(m1A58)-methyltransferase subunit TRM61; tRNA(m1A58)-methyltransferase subunit TRMT61A; tRNA(m1A58)MTase subunit TRM61; tRNA(m1A58)MTase subunit TRMT61A
Gene Aliases: C14orf172; GCD14; Gcd14p; hTRM61; TRM61; TRMT61A
UniProt ID: (Human) Q96FX7
Entrez Gene ID: (Human) 115708
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support