Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: SLRKRKASPEP PDSAEGALKL GEEQQRQQWM ANQSEALKLT MSAG
Highest antigen sequence identity to the following orthologs - mouse 98%, rat 98%.
May contribute to the control of female reproductive function. May play a role in gene transcription by transactivating GNRH1 promoter and repressing PENK promoter.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: enhanced at puberty 1; Enhanced at puberty protein 1; interferon regulatory factor 2 binding protein-like; Interferon regulatory factor 2-binding protein-like; Probable E3 ubiquitin-protein ligase IRF2BPL
Gene Aliases: C14orf4; EAP1; IRF2BPL; KIAA1865; My039
UniProt ID: (Human) Q9H1B7
Entrez Gene ID: (Human) 64207
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support