Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
C16orf87 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: UPF0547 protein C16orf87
Gene Aliases: C16orf87
UniProt ID: (Human) Q6PH81
Entrez Gene ID: (Human) 388272
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support