Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EF-hand calcium-binding domain-containing protein 13; EF-hand domain-containing protein C17orf57
Gene Aliases: C17orf57; EFCAB13
UniProt ID: (Human) Q8IY85
Entrez Gene ID: (Human) 124989
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support