Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
HEATR9 gene ontology annotations related to this gene include hematopoietic progenitor cell differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HEAT repeat containing 9; HEAT repeat-containing protein 9; Protein HEATR9
Gene Aliases: C17orf66; Gm11435; HEATR9; OTTMUSG00000000990; RGD1559875
UniProt ID: (Human) A2RTY3, (Mouse) Q5QNV8
Entrez Gene ID: (Human) 256957, (Rat) 497970, (Mouse) 629303
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support