Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Positive test controls include: U-251MG, LO2, A375, Mouse brain, Mouse skeletal muscle, Mouse testis. The target is usually found in the following locations: Cytoplasm, Nucleus.
Immunogen sequence: MLDSLLALGG LVLLRDSVEW EGRSLLKALV KKSALCGEQV HILGCEVSEE EFREGFDSDI NNRLVYHDFF RDPLNWSKTE EAFPGGPLGA LRAMCKRTDP VPVTIALDSL SWLLLRLPCT TLCQVLHAVS HQDSCPGDSS SVGKVSVLGL LHEELHGPGP VGALSSLAQT EVTLGGTMGQ ASAHILCRRP RQRPTDQTQW FSILPDFSLD LQEGPSVESQ PYSDPHIPPV DPTTHLTFNL HLSKKEREAR DSLILPFQFS SEKQQALLRP RPGQATSHIF YEPDAYDDLD QEDPDDDLDI
C17orf81 belongs to the ELP5 family. C17orf81 may be involved in TP53-mediated transcriptional regulation.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dermal papilla derived protein 6; Dermal papilla-derived protein 6; Dermal papilla-derived protein 6 homolog; Elongator complex protein 5; retinoic acid induced 12; Retinoic acid-induced protein 12; S-phase 2 protein
Gene Aliases: C17orf81; DERP6; ELP5; HSPC002; MST071; MSTP071; Rai12
UniProt ID: (Human) Q8TE02, (Mouse) Q99L85
Entrez Gene ID: (Human) 23587, (Mouse) 54351
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support