Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MICTFLRAV QYTEKLHRSS AKRLLLPYIV LNKACLKTEP SLRCGLQYQK KTLRPRCILG VTQKTIWTQG PSPRKAKEDG SKQVSVHRSQ RGGTAVPTSQ KVKEAGRDFT YLIVVLFGIS ITGGLFYTIF KELFSSSSPS KIYGRALEKC RSHPEVIGVF GESVKGYGEV TRRGRRQHVR FTEYVKDGLK HTCVKFYIEG SEPGKQGTVY AQVKENPGSG EYDFRYIFVE IESYPRRTII IEDNRSQDD (1-248 aa encoded by BC000892 )
Involved in mitochondrial cytochrome c oxidase assembly; mitochondrial respiratory chain complex I assembly; and protein import into mitochondrial matrix. Part of TIM23 mitochondrial import inner membrane translocase complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Mitochondrial import inner membrane translocase subunit Tim21; TIM21-like protein, mitochondrial; translocase of inner mitochondrial membrane 21 homolog
Gene Aliases: 1700034H14Rik; 2700002I20Rik; C18orf55; HSPC154; TIM21; TIMM21
UniProt ID: (Human) Q9BVV7, (Mouse) Q8CCM6
Entrez Gene ID: (Human) 29090, (Mouse) 67105
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support