Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: FENMGLRSP VNGGAGEPPG QLLLQQQRLG RALWHLRLAH HSQRAAFNNN VSVAYECLSA GGRRKRPGLD GRTYSELLRR ICRDGQAPEE VVAPLLRKVQ CRDHEAVPLS VFRAGTLTCF VLLEFVARAG ALFQLLEDSA AAVADRRVGQ AVLDTLEGAL QASDAAAPAR FLEAGSRLGP DSLALALDRA VGGRRPSAPM TREEFLERAA ALFIAKVKPV G (36-290 aa encoded by BC009520)
TPGS1 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: gene trap ROSA b-geo 22; p32; PGs1; PGs1 protein p32; Tubulin polyglutamylase complex subunit 1
Gene Aliases: AV005629; C19orf20; Gm16517; GTRGEO22; PGs1; TPGS1
UniProt ID: (Human) Q6ZTW0, (Mouse) Q99MS8
Entrez Gene ID: (Human) 91978, (Mouse) 110012
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support