Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MASARKASR PMRDVFGDFS DVSLEDSTME EIRNFQISRN LTKIAPGHSR FLKRNQTLDE KHLLLKENPV LGSGPRLASC RPPTTASRIR ANAALMKLAQ LETRIMNRKL QRNLSDTESD SMTADAGLPK RADRILSGGA LELASQNTDK TSQNQARELP VTENNAQNAK VSRFLKKKQA PVENISPEAP AGKERTLQTP KQKEPARTFD SPDSDEEEMK VLLGSLMDSS REKNTNQGFS SANVSEEEER KLFSVPSQLR AFTVPSVELS SAKPSQTSHL PTSLAADRTL HSTRSRADYP QSHVSSDTAS HTPSVSITGA FSNSVSLKMG HVKLVSSPGR SEAETVDEPV S (1-350 aa encoded by BC027869)
The function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ21742; Uncharacterized protein C19orf44; Uncharacterized protein C19orf44 homolog
Gene Aliases: C19orf44
UniProt ID: (Human) Q9H6X5, (Rat) Q6AXP1
Entrez Gene ID: (Human) 84167, (Rat) 290615
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support