Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
The protein encoded by this gene is a DNA binding and apoptosis-inducing protein and is localized in the nucleus. It is also a Rac3-interacting protein which acts as a corepressor for the thyroid hormone receptor. This protein is thought to regulate TRAX/Translin complex formation. Alternate splicing results in multiple transcript variants that encode the same protein. Multiple pseudogenes of this gene are found on chromosome 10.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C1D DNA-binding protein; C1D nuclear receptor co-repressor; hC1D; mC1D; MGC12261; MGC14659; MGC188504; nuclear DNA binding protein; nuclear DNA-binding protein; Nuclear nucleic acid-binding protein C1D; small unique nuclear co-repressor; small unique nuclear receptor co-repressor; Small unique nuclear receptor corepressor; Sun-CoR; sunCoR
Gene Aliases: 1110036E10Rik; AI875855; C1D; hC1D; LRP1; Rrp47; SUN-CoR; SUNCOR
UniProt ID: (Human) Q13901, (Mouse) O35473
Entrez Gene ID: (Human) 10438, (Mouse) 57316
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support