Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: armadillo repeat containing 11; maestro heat-like repeat family member 9; Maestro heat-like repeat-containing protein family member 9
Gene Aliases: 4921528O07Rik; ARMC11; C1orf129; MROH9; RGD1564745
UniProt ID: (Human) Q5TGP6
Entrez Gene ID: (Human) 80133, (Rat) 501864, (Mouse) 78258
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support