Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: SHRFDISAVY PNWKKFSTFT EAPYSTRYST QVSHIGPGTY SSKETCFSKK KLMKEVDTGW AKAQEATRLT QLPHFQYQAI MKEKRLKEQK LGPGSYNLKD FL
Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 71%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ciliary microtubule-associated protein 2; Lymphocyte expansion molecule; uncharacterized protein C1orf177
Gene Aliases: C1orf177; CIMAP2; LEM; LEXM
UniProt ID: (Human) Q3ZCV2
Entrez Gene ID: (Human) 163747
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support