Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: KTTEVPGFD KLVLIIPWVV MQELDRMKEG KLLKRAQHKA IPAVHFINDS LKNQDRKLWG QSIQLASQKH YGLSDENNDD RVLKCCLQHQ ELFPCSFVIL CTDDRNLRNK GLISGVKSLS KEELSAELLR LSLNTDVCHQ PCIPKQQLKA ETTPLKESYK EESTSSGLSI LLESIVSDLE KSLGTGLSSI LETEMKIAFG NLWMEILYLK PPWTLLHLLQ CFKKHWLAVF GLVMEKNLLL TIESLYKNLR KANKAVDFTT VKFLLQDSRS LLHAFSTRSN YDGILPQTFA QVNNLLQTFA EVKTKLKPNS SENTVTKKQE GTSLKNSHN (408-735 aa encoded by BC030781)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ca026; SWT1 RNA endoribonuclease homolog; Transcriptional protein SWT1
Gene Aliases: 1200016B10Rik; C1orf26; HsSwt1; SWT1
UniProt ID: (Human) Q5T5J6, (Mouse) Q9DBQ9
Entrez Gene ID: (Human) 54823, (Mouse) 66875
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support