Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MKAAYLSMFG KEDHKPFGDD EVELFRAVPG LKLKIAGKSL PTEKFAIRKS RRYFSSNPIS LPVPALEMMY IWNGYAVIGK QPKLTDGILE IITKAEEMLE KGPENEYSVD DECLVKLLKG LCLKYLGRVQ EAEENFRSIS ANEKKIKYDH YLIPNALLEL ALLLMEQDRN EEAIKLLESA KQNYKNYSME SRTHFRIQAA TLQAKSSLEN SSRSMVSSVS L
TTC39A is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DEME-6; Differentially expressed in MCF-7 with estradiol protein 6; differentially expressed in MCF7 with estradiol protein 6; testicular tissue protein Li 213; Tetratricopeptide repeat protein 39A; TPR repeat protein 39A
Gene Aliases: C1orf34; DEME-6; KIAA0452; TTC39A
UniProt ID: (Human) Q5SRH9
Entrez Gene ID: (Human) 22996
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support