Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
C1orf53 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Uncharacterized protein C1orf53
Gene Aliases: C1orf53
UniProt ID: (Human) Q5VUE5
Entrez Gene ID: (Human) 388722
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support