Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LGNLVKNTLE KCQRENGFIY FQKIPTEAPQ LELKANYGLV EPIPFEFPPT SVQWTPETLA AFDLTKRPKD DSTKPKPEEE VKPVKEPDIK PQKDTGCYIS
Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 90%.
BROX (BRO1 Domain And CAAX Motif Containing) is a Protein Coding gene with functional area in the extracellular exosome but its exact function is unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BRO1 domain- and CAAX motif-containing protein; BRO1 domain-containing protein BROX; RP11-452F19.1
Gene Aliases: BROFTI; BROX; C1orf58
UniProt ID: (Human) Q5VW32
Entrez Gene ID: (Human) 148362
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support