Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: AGYAHFLNVQG RTLYDVILYG LQEHSEVSGF LLECDSSVQG ALQKHLALYR IRRKVTVEPH PELRVWAVLP SSPEACGAAS LQERAGAAAI LIRD
Highest antigen sequence identity to the following orthologs - mouse 70%, rat 70%.
Involved in the maturation of mitochondrial 4Fe-4S proteins functioning late in the iron-sulfur cluster assembly pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IBA57, iron-sulfur cluster assembly homolog; iron-sulfur cluster assembly factor for biotin synthase- and aconitase-like mitochondrial proteins, with a mass of 57kDa; Iron-sulfur cluster assembly factor homolog; putative transferase C1orf69, mitochondrial; Putative transferase CAF17, mitochondrial
Gene Aliases: C1orf69; IBA57; MMDS3; SPG74
UniProt ID: (Human) Q5T440
Entrez Gene ID: (Human) 200205
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support