Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MARPPVPGSV VVPNWHESAE GKEYLACILR KNRRRVFGLL ERPVLLPPVS IDTASYKIFV SGKSGVGKTA LVAKLAGLEV PVVHHGTTGI QTTVVFWPAK LQASSRVVMF RFEFWDCGES ALKKFDHMLL ACMENTDAFL FLFSFTDRAS FEDLPGQLAR IAGEAPGVVR MVIGSKFDQY MHTDVPERDL TAFRQAWELP LLRVKSVPGR RLADGRTLDG RAGLADVAHI LNGLAEQLWH QDQVAAGLLP NPPESAPE
Potential effector of the planar cell polarity signaling pathway. Plays a role in targeted membrane trafficking most probably at the level of vesicle fusion with membranes. Involved in cilium biogenesis by regulating the transport of cargo proteins to the basal body and to the apical tips of cilia. More generally involved in exocytosis in secretory cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ciliogenesis and planar polarity effector 2; miro domain-containing protein C1orf89; Rem/Rab-Similar GTPase 1; REM2 and RAB-like small GTPase 1; REM2- and Rab-like small GTPase 1
Gene Aliases: C1orf89; CPLANE2; RSG1
UniProt ID: (Human) Q9BU20
Entrez Gene ID: (Human) 79363
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support