Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: SAPSCPQNVN ISGGTFTLSH GWAPGSLLTY SCPQGLYPSP ASRLCKSSGQ WQTPGATRSL SKAVCKPVRC PAPVSFENGI YTPRLGSYPV GGNVSFECED GFILRGSPVR QCRPNGMWDG ETAVCDNGAG HCPNPGISLG AVRTGFRFGH GDKVRYRCSS NLVLTGSSER ECQGNGVWSG TEPICRQPYS YDFPEDVAPA LGTSFSHMLG ATNPTQKTKE SLGRKIQIQR SGHLNLYLLL DCSQSVSEND FLIFKESASL MVDRVRNQES ACSRGLPVLT ISLCLLPLLR TPLTAHLLQE VFSDYTHAM
Component C2 is a serum glycoprotein that functions aspart of the classical pathway of the complement system. Activated C1 cleaves C2 into C2a and C2b. The serine proteinase C2a then combines with complement factor 4b to create the C3 or C5 convertase. Deficiency of C2 has been reported to associated with certain autoimmune diseases and SNPs in this gene have been associated with altered susceptibility to age-related macular degeneration. This gene localizes within the class III region of the MHC on the short arm of chromosome 6. Alternative splicing results in multiple transcript variants encoding distinct isoforms. Additional transcript variants have been described in publications but their full-length sequence has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C3/C5 convertase; Complement C2; complement component C2; DADB-122G4.1
Gene Aliases: ARMD14; C2; CO2
UniProt ID: (Human) P06681
Entrez Gene ID: (Human) 717
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support