Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
The function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: maestro heat-like repeat family member 8; Maestro heat-like repeat-containing protein family member 8; Protein MROH8
Gene Aliases: 4922505G16Rik; C20orf131; C20orf132; dJ621N11.3; dJ621N11.4; MROH8
UniProt ID: (Human) Q9H579
Entrez Gene ID: (Human) 140699, (Mouse) 629499, (Rat) 100360777
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support