Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MQLEYETQIK KMLCSKDSLG LKEELTDEDR EKVAVQEGIN SGTAMSRGLY QPATIFMGRQ MSAILSMRDF STEHKSPQPT KNFSIPDPHS HRQTAQSSNV TDSCVVQTSN DTQCLNKSDN IDGKASLQIG EKTPVTASVL SEEEQTHCLE IGSNTRHGKS NLSEGKKSAE LNSPLRERLS PENRTTDLKC DSSSGSEGEI LTREHIEVEE KRASPPVSPI PVSEYCESEN KWSQEKHSPW EGVSDHLAHR EPKSQKPFRK MQEEEEESWS TSSDLTISIS EDDLILESPE PQPNPGGKME GEDGIEALKL IHAEQERVAL STEKNCILQT LSSPDSEKES STNAPTREPG QTPDSDVPRA QVGQHVATLK EHDNSVKEEA TALLRKALTE ECGRRSAIHS SESSCSLPSI LNDNSGIKEA KPAVWLNSVP TREQEVSSGC GDKSKKENVA ADIPITETEA YQLLKKATLQ DNTNQTENRF QKTDASVSHL SGLNIGSGAF ETKTANKIAS EASFSSSEGS PLSRHENKKK PVINLKSNAL WDESDDSNSE IEAALRPRNH NTDDSDDFYD
Centrosomal protein required for establishing a robust mitotic centrosome architecture that can endure the forces that converge on the centrosomes during spindle formation. Required for stabilizing the expanded pericentriolar material around the centriole.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Centrosomal protein kizuna; Polo-like kinase 1 substrate 1
Gene Aliases: C20orf19; HT013; KIZ; Kizuna; NCRNA00153; PLK1S1; RP69
UniProt ID: (Human) Q2M2Z5
Entrez Gene ID: (Human) 55857
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support