Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: FVNFFGDKTD FHPLMDQFMN DYVEEANREI EKYNQELEQQ EYHDLFELKP
Sequence homology: Cow: 93%; Dog: 93%; Horse: 93%; Human: 100%; Mouse: 86%; Pig: 93%; Rabbit: 93%; Rat: 86%; Yeast: 83%
The function of the C20orf194 gene has not yet been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNAAF9; Dynein axonemal assembly factor 9
Gene Aliases: C20orf194; DNAAF9
UniProt ID: (Human) Q5TEA3
Entrez Gene ID: (Human) 25943
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support