Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MGEAEVGGG GAAGDKGPGE AATSPAEETV VWSPEVEVCL FHAMLGHKPV GVNRHFHMIC IRDKFSQNIG RQVPSKVIWD HLSTMYDMQA LHESEILPFP NPERNFVLPE EIIQEVREGK VMIEEEMKEE MKEDVDPHNG ADDVFSSSGS LGKASEKSSK DKEKNSSDLG CKEGADKRKR SRVTDKVLTA NSNPSSPSAA KRRRT (1-204 aa encoded by BC014235)
C20orf20, also named as MRG/MORF4L-binding protein or Up-regulated in colon cancer 4 (Urcc4), is a 204 amino acid protein, which belongs to the EAF7 family. C20orf20 localizes in the nucleus and is a Component of the NuA4 histone acetyltransferase (HAT) complex which is involved in transcriptional activation of select genes principally by acetylation of nucleosomal histones H4 and H2A. This modification may both alter nucleosome - DNA interactions and promote interaction of the modified histones with other proteins which positively regulate transcription. This complex may be required for the activation of transcriptional programs associated with oncogene and proto-oncogene mediated growth induction, tumor suppressor mediated growth arrest and replicative senescence, apoptosis, and DNA repair. C20orf20 is modified through phosphorylation and acetylation, so the molecular weight of C20orf20 might be 25-30 kDa.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ10914; MRG-binding protein; MRG/MORF4L-binding protein; Up-regulated in colon cancer 4; Urcc4
Gene Aliases: 1600027N09Rik; AW060503; C20orf20; C80444; Eaf7; MRG15BP; MRGBP; URCC4
UniProt ID: (Human) Q9NV56, (Mouse) Q9DAT2
Entrez Gene ID: (Human) 55257, (Mouse) 73247
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support