Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAAANKGNKP RVRSIRFAAG HDAEGSHSHV HFDEKLHDSV VMVTQESDSS FLVKVGFLKI LHRYEITFTL PPVHRLSKDV REAPVPSLHL KLLSVVPVPE GYSVKCEYSA HKEGVLKEEI LLACEGGTGT CVRVTVQARV MDRHHGTPML LDGVKCVGAE LEYDSEHSDW HGFD
Representing about 2% of human DNA, chromosome 20 consists of approximately 63 million bases and 600 genes. Chromosome 20 contains a region with numerous genes expressed in the epididymis, which are thought important for seminal production, and some viewed as potential targets for male contraception. The PRNP gene encoding the prion protein associated with spongiform encephalopathies, like Creutzfeldt-Jakob disease, is found on chromosome 20. Amyotrophic lateral sclerosis, spinal muscular atrophy, ring chromosome 20 epilepsy syndrome and Alagille syndrome are also associated with chromosome 20.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adipose-secreted signaling protein
Gene Aliases: ADISSP; C20orf27
UniProt ID: (Human) Q9GZN8
Entrez Gene ID: (Human) 54976
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support