Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: EKALGKAASH IKSIKNVTEL KLSDNPAWEG DKGNTKGDKH DDLQRARFIC
C20orf43 protein function remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Replication termination factor 2; Replication termination factor 2 domain-containing protein 1; RTF2; UPF0549 protein C20orf43
Gene Aliases: AD-007; C20orf43; CDA05; CDAO5; HSPC164; HSPC169; RTF2; RTFDC1; SHUJUN-3
UniProt ID: (Human) Q9BY42
Entrez Gene ID: (Human) 51507
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support