Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MKLTRKMVLT RAKASELHSV RKLNCWGSRL TDISICQEMP SLEVITLSVN SISTLEPVSR CQRLSELYLR RNRIPSLAEL FYLKGLPRLR VLWLAENPCC GTSPHRYRMT VLRTLPRLQK LDNQAVTEEE LSRALSEGEE ITAAPEREGT GHGGPKLCCT LSSLSSAAET GRDPLDSEEE ATGAQDERGL KPPSRGQFPS LSARDASSSH RGRVSGGPLG AAAASAHCTH CTETVGREHG ASQGPVGREH GASQGLEELC PRGSCVCGSV NAHTRVTRAP HGAVLAPQPL LLSWSVECGP GPCWAEGNRS HVEEVPHTRP QAGLLCSDSP SVPNVLTAIL LLLRELDAEG LEAVQQTVGS RLQALRGEEV QEHAE
Four alternatively spliced transcript variants encoding four different isoforms have been found for this nuclear gene. All isoforms contain leucine-rich repeats. Three of these isoforms are mitochondrial proteins and one of them lacks the target peptide, so is not located in mitochondrion. This gene is down-regulated in Down syndrome (DS) brain, which may represent mitochondrial dysfunction in DS patients.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C21orf-HUMF09G8.5; cDNA A2-YF5; Cilia- and flagella-associated protein 410; cu002; leucine rich repeat containing 76; Leucine-rich repeat-containing protein 76; YF5; YF5/A2
Gene Aliases: C21orf2; CFAP410; LRRC76; YF5/A2
UniProt ID: (Human) O43822
Entrez Gene ID: (Human) 755
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support