Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
The C2CD2 gene, located on chromosome 2, encodes a protein that contains two C2 domains and is involved in cellular signaling and membrane trafficking. The protein has been implicated in various cellular processes, including calcium-dependent phospholipid binding and vesicle transport. Recent studies have shown a critical role for C2CD2 in the regulation of immune responses and cell apoptosis. The protein also interacts with other cellular components to modulate pathways that are essential for cell survival and proliferation. Mutations or dysregulation of C2CD2 have been linked to certain diseases, such as cancers and autoimmune disorders, highlighting its potential as a biomarker and therapeutic target.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C2 calcium-dependent domain containing 2; C2 domain-containing protein 2; TMEM24 related geme; Transmembrane protein 24-like
Gene Aliases: 5730563M15; 5830404H04Rik; AI314146; AI449533; BF168443; C21orf25; C21orf258; C2CD2; Kiaa4045; mKIAA4045; ORF25; RGD735112; TMEM24L
UniProt ID: (Human) Q9Y426, (Mouse) E9Q3C1
Entrez Gene ID: (Human) 25966, (Mouse) 207781, (Rat) 304055
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support