Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
This gene encodes a potential mitochondrial protein that is a member of the DJ-1/PfpI gene family. This protein is overexpressed in fetal Down syndrome brain. Alternate splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Glutamine amidotransferase-like class 1 domain-containing protein 3, mitochondrial; H KNPH; human HES1 protein, homolog to E.coli and zebrafish ES1 protein; Keio novel protein I; testis secretory sperm-binding protein Li 237E
Gene Aliases: C21orf33; ES1; GATD3; GATD3A; GT335; HES1; KNPH; KNPI
UniProt ID: (Human) P0DPI2
Entrez Gene ID: (Human) 8209
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support