Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MVLLHVKRGD ESQFLLQAPG STELEELTVQ VARVYNGRLK VQRLCSEMEE LAEHGIFLPP NMQGLTDDQI EELKLKDEWG EKCVPSGGAV FKKDDIGRRN GQAPNEKMKQ VLKKTIEEAK AIISKKQVEA GVCVTMEMVK DALDQLRGAV MIVYPMGLPP YDPIRMEFEN KEDLSGTQAG LNVIKEAEAQ LWWAAKELRR TKKLSDYVGK NEKTKIIAKI QQRGQGAPAR EPIISSEEQK QLMLYYHRRQ EELKRLEEND DDAYLNSPWA DNTALKRHFH GVKDIKWRPR
The function of the C21orf59 protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cilia- and flagella-associated protein 298; prostate cancer upregulated protein 1; Protein kurly homolog
Gene Aliases: C21orf48; C21orf59; CFAP298; CILD26; FBB18; Kur
UniProt ID: (Human) P57076
Entrez Gene ID: (Human) 56683
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support